missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Akirin2 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20608 can be used with blocking peptide PEP-0728.
The highly conserved, nuclear-localized Akirin1 and Akirin2 proteins critically regulate the transcription of NF-kappa-B-dependent genes and are required for defense against Gram-negative bacteria in the immune deficiency and NF-kappa-B pathways. Akirin1 is dispensable in the mouse, and neither knockout mice nor cells derived from them have obvious distinctive phenotypes. In contrast, Akirin2 is required for development in the mouse and knockout of both Akirin homologs in mice show that Akirin2 is required downstream of toll-like receptor (TLR), TNF-a and IL-1-b signaling, and for the production of IL-6. Akirin2 is functionally closer to the single gene in Drosophila, as the homozygous null D. melanogaster Akirin mutants show a similar, mid-to-early embryonic death.
Specifications
Specifications
| Antigen | Akirin2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | AKIRIN2 |
| Gene Accession No. | B1AXD8, Q25C79, Q53H80 |
| Gene Alias | 14-3-3 beta; 2700059D21Rik; AA114675; AA522011; akirin 2; Akirin2; Akirin-2; akirin2 {ECO:0000250; AU019887; C6orf166; dJ486L4.2; EMBL:BAE81786.1}; FBI1; Fourteen-three-three beta interactant 1; fourteen-three-three beta interactant 1 {ECO:0000312; fourteen-three-three beta interacting-protein 1; RGD1307791; UniProtKB:Q53H80} |
| Gene Symbols | AKIRIN2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?