missing translation for 'onlineSavingsMsg'
Learn More

Akirin2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18409916 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409916 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409916 Supplier Novus Biologicals Supplier No. NBP176347AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Akirin2 Polyclonal antibody specifically detects Akirin2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Akirin2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias akirin 2, akirin-2, C6orf166, chromosome 6 open reading frame 166, dJ486L4.2, FBI1, FLJ10342, fourteen-three-three beta interactant 1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the center of the human Akirin2. The immunogen is located within amino acids 80 - 130 of Akirin2. Amino Acid Squence: ETSFQQTDPCCTSDA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 55122
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.