missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKIP Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00054998-B01P-30ug
This item is not returnable.
View return policy
Description
AKIP Polyclonal antibody specifically detects AKIP in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| AKIP | |
| Polyclonal | |
| Western Blot 1:500, Immunocytochemistry/ Immunofluorescence | |
| NP_060370.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| PBS (pH 7.4) | |
| AIPAKIPaurora kinase A-interacting protein, AURKA-interacting protein, aurora kinase A interacting protein 1, aurora-A kinase interacting protein, FLJ20608 | |
| AURKAIP1 (NP_060370.1, 1 a.a. - 199 a.a.) full-length human protein. MLLGRLTSQLLRAVPWAGGRPPWPVSGVLGSRVCGPLYSTSPAGPGRAASLPRKGAQLELEEMLVPRKMSVSPLESWLTARCFLPRLDTGTAGTVAPPQSYQCPPSQIGEGAEQGDEGVADAPQIQCKNVLKIRRRKMNHHKYRKLVKKTRFLRRKVQEGRLRRKQIKFEKDLRRIWLKAGLKEAPEGWQTPKIYLRGK | |
| 30 μg | |
| Cell Cycle and Replication | |
| 54998 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction