missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27554-1-AP-150UL
This item is not returnable.
View return policy
Description
Involved in maintaining the homeostasis of cellular nucleotides by catalyzing the interconversion of nucleoside phosphates. Has both nucleoside monophosphate and diphosphate kinase activities. Catalyzes the phosphorylation of AMP, dAMP, CMP and dCMP with ATP as phosphate donor and of CMP with GTP as phosphate donor. Also catalyzes the production of ATP, CTP, GTP, UTP, dATP, dCTP, dGTP and TTP from the corresponding diphosphate substrates with either ATP or GTP as phosphate donor. Shows substrate preference of CDP > UDP > ADP > GDP > TDP.Specifications
| AKD2 | |
| Polyclonal | |
| Unconjugated | |
| AK9 | |
| Adenylate kinase 9, AK 9, AK9, AKD1, AKD2, C6orf199, C6orf224, RP1 70A9.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221264 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5TCS8 | |
| AK9 | |
| AKD2 Fusion Protein Ag25805 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction