missing translation for 'onlineSavingsMsg'
Learn More

AKAP95/AKAP8 Antibody, Novus Biologicals™

Product Code. 18325900 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325900 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325900 Supplier Novus Biologicals Supplier No. NBP286972

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AKAP95/AKAP8 Polyclonal specifically detects AKAP95/AKAP8 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP95/AKAP8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias A kinase (PRKA) anchor protein 8, AKAP-8, AKAP-95 AKAP 95, A-kinase anchor protein 8, A-kinase anchor protein 95 kDa, A-kinase anchor protein, 95kDa, DKFZp586B1222
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human AKAP95/AKAP8
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10270
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.