missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-89166
This item is not returnable.
View return policy
Description
AKAP9 Polyclonal specifically detects AKAP9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| AKAP9 | |
| Polyclonal | |
| Western Blot Reported in scientific literature (PMID 25366663)., Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| A kinase (PRKA) anchor protein (yotiao) 9, AKAP 350, AKAP 450, AKAP350A-kinase anchor protein 450 kDa, AKAP450A-kinase anchor protein 350 kDa, AKAP-9, Centrosome- and Golgi-localized PKN-associated protein, CG-NAPAKAP 120-like protein, hgAKAP 350, HYPERION, KIAA0803A-kinase anchor protein 9, MU-RMS-40.16A, PRKA9AKAP9-BRAF fusion protein, Protein hyperion, protein kinase A anchoring protein 9, Protein kinase A-anchoring protein 9, Protein yotiao, YOTIAOkinase N-associated protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10142 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| AKAP9 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KLLEAISETSSQLEHAKVTQTELMRESFRQKQEATESLKCQEELRERLHEESRAREQLAVELSKAEGVIDGYADEKTLFERQIQEKTDIIDRLEQELLCASNRLQELEAEQQQIQEERELLSRQKEAMKAEAGPVEQQLLQETE | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction