Learn More
AKAP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12591-1-AP-150UL
 View more versions of this product
Product Code. 16800384
  • £349.00 / 150µL
Description
This antibody can recognize all the isoforms of AKAP7.
This gene encodes a member of the A-kinase anchoring protein family, a group of functionally related proteins that bind to a regulatory subunit of cAMP-dependent protein kinase A and target the enzyme to specific subcellular compartments. AKAPs have a common RII-binding domain, but contain different targeting motifs responsible for directing PKA to distinct intracellular locations. Three alternatively spliced transcript variants encoding different isoforms have been described. Additional variants exist, but their full-length natures have not been determined.
Specifications
Specifications
AKAP7
Polyclonal
Unconjugated
AKAP7
A kinase anchor protein 18 kDa, AKAP 18, AKAP 7 isoforms alpha and beta, AKAP18, AKAP7
Rabbit
Antigen Affinity Chromatography
RUO
361458, 432442, 9465
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43687, O55074, Q6JP77
Akap7
AKAP7 Fusion Protein Ag3297
150 μL
Primary
Human, Mouse, Rat