missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-53116
This item is not returnable.
View return policy
Description
AKAP7 Polyclonal specifically detects AKAP7 in Human samples. It is validated for Western Blot.
Specifications
| AKAP7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| A kinase (PRKA) anchor protein 7, AKAP 18, AKAP15, AKAP18A-kinase anchor protein 7 isoform alpha, AKAP-7 isoform gamma, AKAP-7 isoforms alpha and beta, A-kinase anchor protein 18 kDa, A-kinase anchor protein 7, A-kinase anchor protein 7 isoform gamma, A-kinase anchor protein 7 isoforms alpha and beta, A-kinase anchor protein 9 kDa, A-kinase anchor protein, 18-kD, A-kinase anchoring protein 18, protein kinase A anchoring protein 7, Protein kinase A-anchoring protein 7 isoform gamma, Protein kinase A-anchoring protein 7 isoforms alpha/beta | |
| Rabbit | |
| 37 kDa | |
| 100 μL | |
| Stem Cell Markers | |
| 9465 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Rabbit | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9P0M2 | |
| AKAP7 | |
| Synthetic peptides corresponding to AKAP7(A kinase (PRKA) anchor protein 7) The peptide sequence was selected from the middle region of AKAP7. Peptide sequence MKLSKSPWLRKNGVKKIDPDLYEKFISHRFGEEILYRIDLCSMLKKKQSN. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction