missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38590
This item is not returnable.
View return policy
Description
AKAP6 Polyclonal specifically detects AKAP6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AKAP6 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Q13023 | |
| AKAP6 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SHSATKSALIQKLMQDIQHQDNYEAIWEKIEGFVNKLDEFIQWLNEAMETTENWTPPKAEMDDLKLYLETHLSFKLNVDSHCALKEAVEEEGHQLLELIASHKAGLKDMLRMIASQWKELQRQIKRQHSWILRALDTIKA | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| A kinase (PRKA) anchor protein 6, ADAP6A-kinase anchor protein 6, AKAP100A-kinase anchor protein 100 kDa, AKAP-6, KIAA0311AKAP 100, mAKAPMGC165020, PRKA6ADAP100, protein kinase A anchoring protein 6, Protein kinase A-anchoring protein 6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9472 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction