missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP5 Polyclonal Antibody

AKAP5 Polyclonal Antibody
Brand: Invitrogen PA5101095
This item is not returnable.
View return policy
Description
AKAP5 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| AKAP5 | |
| Polyclonal | |
| Unconjugated | |
| AKAP5 | |
| 3526401B18Rik; A kinase (PRKA) anchor protein 5; AKAP 150; AKAP 5; AKAP 79; AKAP150; AKAP5; AKAP-5; AKAP75; AKAP79; A-kinase anchor protein 150 kDa; A-kinase anchor protein 5; A-kinase anchor protein 79 kDa; A-kinase anchoring protein 5; A-kinase anchoring protein 75/79; BB098886; cAMP-dependent protein kinase regulatory subunit II high affinity-binding protein; Gm258; H21; P150; RII-B-binding protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 238276, 9495 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D3YVF0, P24588 | |
| AKAP5 | |
| A synthesized peptide derived from human AKAP5(Accession P24588), corresponding to amino acid residues M1-F37. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction