missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538594
This item is not returnable.
View return policy
Description
AKAPs (A-kinase anchoring proteins), as their name implies, are a family of scaffolding proteins that bind regulatory subunits of Protein Kinase A (PKA) thus localizing PKA activity to distinct regions of the cell. Beyond a common amphipathic alpha helix that is responsible for recruiting the PKA regulatory subunit (RI alpha, RII alpha, RI beta, or RII beta), individual AKAPs contain additional domains responsible for the recrutiment of additional signaling proteins (phosphodiesterases, phosphatases, cytoskeletal components, other kinase, etc.) or restricting AKAP to a specific subcellular location. AKAP5 (also known as P75, AKAP75, or AKAP79) is predomitly expressed in neuronal tissues and cells where it serves to localize type II PKA to post-synaptic densities. AKAP5 specifically binds to the regulatory subunit of PKAII beta, anchoring the enzyme to the plasma membrane and sites of cytoskeletal/membrane junctions. The other binding domains of AKAP5 have been shown to interact with calmodulin, PP2B, and calcineurin suggesting that AKAP5 may act to coordinate the cAMP- and Ca2+-sensing pathways in various cell types.
Specifications
| AKAP5 | |
| Polyclonal | |
| Unconjugated | |
| AKAP5 | |
| A kinase (PRKA) anchor protein 5, A kinase anchor protein 5, A kinase anchor protein 79kDa, A-kinase anchor protein 5, A-kinase anchor protein 79 kDa, AKAP 5, AKAP 79, AKAP-5, AKAP5, AKAP75, AKAP79 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9495 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P24588 | |
| AKAP5 | |
| A synthetic peptide derived from the N-terminal region of human AKAP5 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion