missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £368.00
Specifications
| Antigen | AKAP5 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638172
|
Novus Biologicals
NBP2-92620-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609071
|
Novus Biologicals
NBP2-92620-0.1ml |
0.1 mL |
£368.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKAP5 Polyclonal antibody specifically detects AKAP5 in Human, Mouse samples. It is validated for Western BlotSpecifications
| AKAP5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 9495 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| A kinase (PRKA) anchor protein 5, AKAP 79, AKAP7979kDa, A-kinase anchor protein 79 kDa, A-kinase anchoring protein 75/79, cAMP-dependent protein kinase regulatory subunit II high affinity bindingprotein, cAMP-dependent protein kinase regulatory subunit II high affinity-bindingprotein, H21 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human AKAP5 (NP_004848.3). METTISEIHVENKDEKRSAEGSPGAERQKEKASMLCFKRRKKAAKALKPKAGSEAADVARKCPQEAGASDQPEPTRGAWASLKRLVTRRKRSESSKQQKPLEGEMQPAINAEDADLSKKKAKSRLKIPCIKFPRGPKRSNHSKIIEDSDCSIKVQEEAEILDIQTQTPLNDQATKAKSTQDLSEGISRKDGDEVCESNVSNSTTSGEKVISVELGLDNGHSAIQTGTLILEEIETIKEKQDVQPQQASPLETSETDHQQPVLSDVPPLPA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title