missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24986-1-AP-150UL
This item is not returnable.
View return policy
Description
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. AKAP4, also known as hAKAP82, is a major sperm fibrous sheath protein that localizes to the entire length of the flagellum of human sperm. It has been suggested that AKAP4 plays multiple roles in sperm motility and regulation of signal transduction pathways. AKAP4 has also been postulated to be a potential biomarker for breast cancer.Specifications
| AKAP4 | |
| Polyclonal | |
| Unconjugated | |
| AKAP4 | |
| A kinase anchor protein 4, A kinase anchor protein 82 kDa, AKAP 4, AKAP 82, AKAP4, AKAP82, CT99, FSC1, hAKAP82, HI, p82, PRKA4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8852 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5JQC9 | |
| AKAP4 | |
| AKAP4 Fusion Protein Ag21762 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction