missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKAP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550314
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total AKAP4 protein.
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. AKAP4, also known as hAKAP82, is a major sperm fibrous sheath protein that localizes to the entire length of the flagellum of human sperm. It has been suggested that AKAP4 plays multiple roles in sperm motility and regulation of signal transduction pathways. AKAP4 has also been postulated to be a potential biomarker for breast cancer.
Specifications
| AKAP4 | |
| Polyclonal | |
| Unconjugated | |
| AKAP4 | |
| 75 kDa fibrous sheath protein; A kinase (PRKA) anchor protein 4; AKAP 82; AKAP4; AKAP-4; akap4 {ECO:0000312; AKAP82; A-kinase anchor protein 4; A-kinase anchor protein 82 kDa; A-kinase anchoring protein 4; cancer/testis antigen 99; CT99; Fsc1; hAKAP82; HI; major sperm fibrous sheath protein; mAKAP82; p82; PRKA4; protein kinase A anchoring protein 4; protein kinase A-anchoring protein 4; RGD:620828}; testis-specific gene HI | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8852 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q5JQC9 | |
| AKAP4 | |
| Fusion protein corresponding to residues near the C terminal of human A kinase (PRKA) anchor protein 4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction