missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£328.00 - £470.00
Specifications
| Antigen | AKAP4 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18435561
|
Novus Biologicals
NBP1-89165-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18266146
|
Novus Biologicals
NBP1-89165 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKAP4 Polyclonal specifically detects AKAP4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AKAP4 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 8852 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SALKLIQYHLTQQTKGKDTCEEDCPGSTMGYMAQSTQYEKCGGGQSAKALSVKQLESHRAPGPSTCQKENQHLDSQKMDMSNIVLMLIQKLLNENPFKCEDPCEGENKCSEPRASKAASMSNRSDKAEEQCQEHQELDCTSGMKQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| A kinase (PRKA) anchor protein 4, AKAP-4, AKAP82AKAP 82, A-kinase anchor protein 4, A-kinase anchor protein 82 kDa, cancer/testis antigen 99, CT99, hAKAP82Fsc1, HIFSC1, Major sperm fibrous sheath protein, p82, PRKA4, protein kinase A anchoring protein 4, Protein kinase A-anchoring protein 4, testis-specific gene HI | |
| AKAP4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title