missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575842
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The type II cAMP-dependent protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins, designated A-kinase anchoring proteins (AKAP). AKAP 3, also known as AKAP 110, FSP95, PRKA3 and SOB1, binds both PKA and PDE4A and functions as a scaffolding protein in spermatozoa to regulate local cAMP concentrations and modulate sperm functions. Expression of AKAP 3 in normal tissues is restricted to the testis, where bicarbonate stimulates tyrosine phosphorylation of AKAP 3, thereby increasing its recruitment of PKA. AKAP-3 also exhibits high expression in patients with epithelial ovarian cancer (EOC). It demonstrates tumor-restricted expression and appears to be associated with worse overall survival, which make AKAP 3 a potential target for antigen-specific immunotherapy in EOC.
Specifications
| AKAP3 | |
| Polyclonal | |
| Unconjugated | |
| AKAP3 | |
| A kinase (PRKA) anchor protein 3; AKAP 110; AKAP110; Akap3; AKAP-3; A-kinase anchor protein 110 kDa; A-kinase anchor protein 3; A-kinase anchor protein, 110kDa; A-kinase anchoring protein 3; Cancer/testis antigen 82; CT82; epididymis luminal protein 159; Fibrous Sheath Protein of 95 kDa; fibrousheathin 1; Fibrousheathin I; fibrousheathin-1; FSP95; HEL159; PRKA3; protein kinase A binding protein AKAP 110; Protein kinase A-anchoring protein 3; SOB1; Sperm oocyte-binding protein; sperm oocyte-binding protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10566, 11642, 312720 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75969, O88987 | |
| AKAP3 | |
| Synthetic peptide corresponding to residues in Human AKAP3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction