missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101277
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total AKAP2.
Binds to regulatory subunit (RII) of protein kinase A. May be involved in establishing polarity in signaling systems or in integrating PKA-RII isoforms with downstream effectors to capture, amplify and focus diffuse, trans-cellular signals carried by cAMP. [UniProt].
Specifications
| AKAP2 | |
| Polyclonal | |
| Unconjugated | |
| AKAP2 | |
| A kinase (PRKA) anchor protein 2; AA959716; AI649048; AKAP expressed in kidney and lung; AKAP2; AKAP-2; AKAPKL; AKAP-KL; A-kinase anchor protein 2; A-kinase anchoring protein 2; B230340M18Rik; KL2B; MISP2; PALM2-AKAP2; PRKA2; protein kinase A anchoring protein 2; protein kinase A2; protein kinase A-anchoring protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11217 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y2D5 | |
| AKAP2 | |
| A synthesized peptide derived from human AKAP2(Accession Q9Y2D5), corresponding to amino acid residues S78-A128. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction