missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP2 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61604G
This item is not returnable.
View return policy
Description
AKAP2 Polyclonal antibody specifically detects AKAP2 in Human, Sheep samples. It is validated for Western Blot
Specifications
| AKAP2 | |
| Polyclonal | |
| Western Blot | |
| A kinase (PRKA) anchor protein 2, AKAP-2, FLJ53197, protein kinase A2 | |
| The immunogen recognized by this antibody maps to a region between residue 525 and 575 of human A kinase (PRKA) anchor protein 2 using the numbering given in entry Q9Y2D5.2 (GeneID 11217). | |
| 0.1 mL | |
| Protein Kinase | |
| 445815 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction