missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKAP14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599280
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total AKAP14.
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The protein anchors PKA in ciliary axonemes and, in this way, may play a role in regulating ciliary beat frequency. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| AKAP14 | |
| Polyclonal | |
| Unconjugated | |
| AKAP14 | |
| A kinase (PRKA) anchor protein 14; AKAP 28; AKAP14; AKAP-14; AKAP28; A-kinase anchor protein 14; A-kinase anchor protein 28 kDa; A-kinase anchoring protein 14; A-kinase anchoring protein 28; PRKA14; Protein kinase A-anchoring protein 14; TAKAP-1.2; Takap80; Testis-specific A-kinase-anchoring protein TAKAP-80; Testis-specific A-kinase-anchoring-protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 158798 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86UN6 | |
| AKAP14 | |
| A synthesized peptide derived from human AKAP14(Accession Q86UN6), corresponding to amino acid residues M1-D34. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction