missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP13, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00011214-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MSSSKKSKEKEKEKDKIKEKEKDSKDKEKDKKTVNGHTFSSIPVVGPISCSQCMKPFTNKDAYTCANCSAFVHKGCRESLASCAKVKMKQPKGSLQAHDTSSLPTVIMRNKPSQPKERPRSAVLLVDETATTPIFANRRSQQSVSLSKSVSIQNITGVGNDENMSNTWKFLSHSTDSLNKISKVNESTESLTDEGVGTDMNEGQLLGDFEIESKQLEAESWSRIIDSKFLKQQKKDVVKRQEVIYELMQTEFHHVRTLKIMSGVYSQGMMADLLFEQQMVEKLFPCLDELISIHSQFFQRILERKKESLVDKSEKNFLIKRIGDVLVNQFSGENAERLKKTYGKFCGQHNQSVNYFKDLYAKDKRFQAFVKKKMSSSVVRRLGIPECILLVTQRITKYPVLFQRILQCTKDNEVEQEDLAQSLSLVKDVIGAVDSKVASYEKKVRLNEIYTKTDSKSIMRMKSGQMFAKEDLKRKKLVRDGSVFLKNAAGRLKEVQAVLLTDILVFLQEKDQKYIFASLDQKSTVISLKKLIVREVAHEEKGLFLISMGMTDPEMVEVHASSKEERNSWIQIIQDTINTLNRDEDEGIPSENEEEKKMLDTRARELKEQLHQKDQKILLLLEEKEMIFRDMAECSTPLPEDCSPTHSPRVLFRSNTEEALKGGPLMKSAINEVEILQGLVSGNLGGTLGPTVSSPIEQDVVGPVSLPRRAETFGGFDSHQMNASKGGEKEEGDDGQDLRRTESDSGLKKGGNANLVFMLKRNSEQVVQSVVHLYELLSALQGVVLQQDSYIEDQKLVLSERALTRSLSRPSSLIEQEKQRSLEKQRQDLANLQKQQAQYLEEKRRREREWEARERELREREALLAQREEEVQQGQQDLEKEREELQQKKGTYQYDLERLRAAQKQLEREQEQLRREAERLSQRQTERDLCQVSHPHTKLMRIPSFFPSPEEPPSPSAPSIAKSGSLDSELSVSPKRNSISRTHKDKGPFHILSSTSQTNKGPEGQSQAPASTSASTRLFGLTKPKEKKEKKKKNKTSRSQPGDGPASEVSAEGEEIFCSpecifications
| AKAP13 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_144767 | |
| AKAP13 | |
| AKAP13 (NP_658913.1, 1 a.a. ∼ 1058 a.a) full-length human protein. | |
| RUO | |
| 11214 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| A kinase (PRKA) anchor protein 13 | |
| AKAP-Lbc/ARHGEF13/BRX/FLJ11952/FLJ43341/HA-3/Ht31/LBC/PROTO-LB/PROTO-LBC/c-lbc | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction