missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12356-1-AP-150UL
This item is not returnable.
View return policy
Description
The A-kinase anchor proteins are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein interacts with both the type I and type II regulatory subunits of PKA; therefore, it is a dual-specific AKAP. This protein is highly enriched in mitochondria. It contains RGS domains, in addition to a PKA-RII subunit-binding domain. The mitochondrial localization and the presence of RGS domains may have important implications for the function of this protein in PKA and G protein signal transduction.Specifications
| AKAP10 | |
| Polyclonal | |
| Unconjugated | |
| AKAP10 | |
| AKAP 10, AKAP10, D AKAP 2, D AKAP2, PRKA10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11216, 360540, 56697 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43572, O88845 | |
| Akap10 | |
| AKAP10 Fusion Protein Ag3020 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction