missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98506-100ul
This item is not returnable.
View return policy
Description
AKAP10 Polyclonal antibody specifically detects AKAP10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| AKAP10 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| A kinase (PRKA) anchor protein 10, AKAP-10, A-kinase anchor protein 10, mitochondrial, D-AKAP2, D-AKAP-2, Dual specificity A kinase-anchoring protein 2, dual-specificity A-kinase anchoring protein 2, MGC9414, mitochondrial A kinase PPKA anchor protein 10, PRKA10a kinase anchor protein 10, mitochondrial, protein kinase A anchoring protein 10, Protein kinase A-anchoring protein 10 | |
| Produced in rabbits immunized with E. coli-derived Human AKAP10 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 11216 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction