missing translation for 'onlineSavingsMsg'
Learn More

AKAP1 Antibody, Novus Biologicals™

Product Code. 18319918 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319918 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319918 Supplier Novus Biologicals Supplier No. H00008165B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

AKAP1 Polyclonal antibody specifically detects AKAP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. AAH00729.1
Gene Alias A kinase (PRKA) anchor protein 1, a kinase anchor protein 1, mitochondrial, AKAP121, AKAP149AKAP 149, AKAP84, A-kinase anchor protein 1, mitochondrial, A-kinase anchor protein 149 kDa, A-kinase anchor protein, 149kD, D-AKAP1, D-AKAP-1, Dual specificity A-kinase-anchoring protein 1, dual-specificity A-kinase anchoring protein 1, MGC1807, PRKA1, protein kinase A anchoring protein 1, protein kinase A1, Protein kinase A-anchoring protein 1, protein kinase anchoring protein 1, S-AKAP84, SAKAP84AKAP, Spermatid A-kinase anchor protein 84
Host Species Mouse
Immunogen AKAP1 (AAH00729.1, 1 a.a. - 903 a.a.) full-length human protein. MAIQFRSLFPLALPGMLALLGWWWFFSRKKGHVSSHDEQQVEAGAVQLRADPAIKEPLPVEDVCPKVVSTPPSVTEPPEKELSTVSKLPAEPPALLQTHPPCRRSESSGILPNTTDMRLRPGTRRDDSTKLELALTGGEAKSIPLECPLSSPKGVLFSSKSAEVCKQDSPFSRVPRKVQPGYPVVPAEKRSSGERARETGGAEGTGDAVLGEKVLEEALLSREHVLELENSKGPSLASLEGEEDKGKSSSSQVVGPVQEEEYVAEKLPSRFIESAHTELAKDDAAPAPPVADAKAQDRGVEGELGNEESLDRNEEGLDRNEEGLDRNEESLDRNEEGLDRNEEIKRAAFQIISQVISEATEQVLATTVGKVAGRVCQASQLQGQKEESCVPVHQKTVLGPDTAEPATAEAAVAPPDAGLPLPGLPAEGSPPPKTYVSCLKSLLSSPTKDSKPNISAHHISLASCLALTTPSEELPDRAGILVEDATCVTCMSDSSQSVPLVASPGHCSDSFSTSGLEDSCTETSSSPRDKAITPPLPESTVPFSNGVLKGELSDLGAEDGWTMDAEADHSGGSDRNSMDSVDSCCSLKKTESFQNAQAGSNPKKVDLIIWEIEVPKHLVGRLIGKQGRYVSFLKQTSGAKIYISTLPYTQSVQICHIEGSQHHVDKALNLIGKKFKELNLTNIYAPPLPSLALPSLPMTSWLMLPDGITVEVIVVNQVNAGHLFVQQHTHPTFHALRSLDQQMYLCYSQPGIPTLPTPVEITVICAAPGADGAWWRAQVVASYEETNEVEIRYVHYGGYKRVKVDVLRQIRSDFVTLPFQGAEVLLDSVMPLSDDDQFSPEADAAMSEMTGNTALLAQVTSYSPTGLPLIQLWSVVGDEVVLINRSLVERGLAQWVDSYYTSL
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8165
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.