missing translation for 'onlineSavingsMsg'
Learn More

AKAIN1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18340170 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340170 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340170 Supplier Novus Biologicals Supplier No. NBP281698

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AKAIN1 Polyclonal specifically detects AKAIN1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKAIN1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2 μg/mL
Formulation PBS
Gene Alias C18orf42, C18orf42 chromosome 18 open reading frame 42
Gene Symbols C18orf42
Host Species Rabbit
Immunogen AKAIN1 antibody was raised against a 16 amino acid peptide from near the center of human AKAIN1. The immunogen is located within the last 50 amino acids of AKAIN1. Amino Acid Squence: VSQESQRREERISDNR
Molecular Weight of Antigen 12 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 642597
Test Specificity AKAIN1 antibody is human specific.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.