missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92176-0.1ml
This item is not returnable.
View return policy
Description
AK7 Polyclonal antibody specifically detects AK7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| AK7 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| adenylate kinase 7, EC 2.7.4.3, FLJ32864, putative adenylate kinase 7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human AK7 (NP_689540.2). MAEEEETAALTEKVIRTQRVFINLLDSYSSGNIGKFLSNCVVGASLEEITEEEEEEDENKSAMLEASSTKVKEGTFQIVGTLSKPDSPRPDFAVETYSAISREDLLMRLLECDVIIYNITESSQQMEEAIWAVSALSEEVSHFEKRKLFILLSTVMTWARSKALDPEDSEVPFTEEDYRRRKSHPNFLDHINAEKMVLKF | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 122481 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction