missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP3-35549-20ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Beschreibung
AK5 Polyclonal antibody specifically detects AK5 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
| AK5 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| adenylate kinase 5, adenylate kinase 6, adenylate kinase isoenzyme 5, AK 5, AK6, ATP-AMP transphosphorylase 5, EC 2.7.4.3, MGC33326 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 150-370 of human AK5 (NP_777283.1).,, Sequence:, LKIAERYGFQYISVGELLRKKIHSTSSNRKWSLIAKIITTGELAPQETTITEIKQKLMQIPDEEGIVIDGFPRDVAQALSFEDQICTPDLVVFLACANQRLKERLLKRAEQQGRPDDNVKATQRRLMNFKQNAAPLVKYFQEKGLIMTFDADRDEDEVFYDISMAVDNKLFPNKEAAAGSSDLDPSMILDTGEIIDTGSDYEDQGDDQLNVFGEDTMGGFM | |
| 20 μL | |
| metabolism | |
| 26289 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction