missing translation for 'onlineSavingsMsg'
Learn More

AK3L1 Antibody (010), DyLight 488, Novus Biologicals™

Product Code. 18363180 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363180 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363180 Supplier Novus Biologicals Supplier No. NBP290093G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

AK3L1 Monoclonal specifically detects AK3L1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AK3L1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 10
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias adenylate kinase 3, Adenylate kinase 3-like, adenylate kinase 3-like 1, adenylate kinase 4, AK3adenylate kinase isoenzyme 4, mitochondrial, AK3L1, AK3L2, ATP-AMP transphosphorylase, EC 2.7.4, EC 2.7.4.3, GTP:AMP phosphotransferase, MGC166959, mitochondrial adenylate kinase-3, nucleoside-triphosphate-adenylate kinase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human AK3L1 (P27144-1; Ala 2-Tyr 223).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 205
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.