missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11014-1-AP-20UL
This item is not returnable.
View return policy
Description
Adenylate kinases are involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate groups among adenine nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. Isozyme 2 is localized in the mitochondrial intermembrane space and may play a role in apoptosis.Specifications
| AK2 | |
| Polyclonal | |
| Unconjugated | |
| AK2 | |
| adenylate kinase 2, ADK2, AK 2, AK2, ATP AMP transphosphorylase 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11637, 204, 24184, 321793 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P29410, P54819, Q1L8L9, Q9WTP6 | |
| ak2 | |
| AK2 Fusion Protein Ag1464 | |
| 20 μL | |
| Primary | |
| Mouse, Zebrafish, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction