missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11014-1-AP-150UL
This item is not returnable.
View return policy
Description
Adenylate kinases are involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate groups among adenine nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. Isozyme 2 is localized in the mitochondrial intermembrane space and may play a role in apoptosis.Specifications
| AK2 | |
| Polyclonal | |
| Unconjugated | |
| AK2 | |
| adenylate kinase 2, ADK2, AK 2, AK2, ATP AMP transphosphorylase 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11637, 204, 24184, 321793 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P29410, P54819, Q1L8L9, Q9WTP6 | |
| ak2 | |
| AK2 Fusion Protein Ag1464 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur