missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AK2 Monoclonal antibody specifically detects AK2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Dilution | Western Blot, ELISA, Immunohistochemistry-Paraffin |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human AK2 (Uniprot#: P54819-1; Met1-Ile239). |
| Primary or Secondary | Primary |
| Content And Storage | Store at 4C in the dark. |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?