missing translation for 'onlineSavingsMsg'
Learn More

AK-7, MedChemExpress

Product Code. 16427146
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427146 100 mg 100mg
16413167 50 mg 50mg
16407138 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16427146 Supplier Medchem Express Supplier No. HY16691

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AK-7 is a selective cell- and brain-permeable SIRT2 inhibitor, with an IC50 of 15.5 μM.

AK-7 is a selective cell- and brain-permeable SIRT2 inhibitor, with an IC50 of 15.5 μM.

Specifications

Chemical Name or Material AK-7
CAS 420831-40-9
Color White
Purity Grade Notes Research
Molecular Formula C19H21BrN2O3S
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (114.32 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(N1CCCCCC1)(C2=CC(C(NC3=CC(Br)=CC=C3)=O)=CC=C2)=O
Molecular Weight (g/mol) 437.35
Formula Weight 437.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.