missing translation for 'onlineSavingsMsg'
Learn More

AK-1, MedChemExpress

Product Code. 16483378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483378 10 mg 10mg
16469906 50 mg 50mg
16446107 5 mg 5mg
16432746 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483378 Supplier Medchem Express Supplier No. HY101465
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AK-1 is a potent, specific and cell-permeable SIRT2 inhibitor, with an IC50 of 12.5 μM.

AK-1 is a potent, specific and cell-permeable SIRT2 inhibitor, with an IC50 of 12.5 μM.

Specifications

Chemical Name or Material AK-1
CAS 330461-64-8
Color White
Purity Grade Notes Research
Molecular Formula C19H21N3O5S
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (123.93 mM)
SMILES O=C(NC1=CC=CC([N+]([O-])=O)=C1)C2=CC=CC(S(=O)(N3CCCCCC3)=O)=C2
Molecular Weight (g/mol) 403.45
Formula Weight 403.45
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.