missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ajmaline, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Ajmaline |
| CAS | 4360-12-7 |
| Molecular Formula | C20 H26 N2 O2 |
| Quantity | 250 mg |
| Synonym | Ajmalan-17,21-diol, (17R,21α)-, Ajmaline (6CI,7CI,8CI), 5H-6,10:11,12a-Dimethanoindolo[3,2-b]quinolizine, ajmalan-17,21-diol deriv., (+)-Ajmaline, Ajmalin, Cardiorythmine, Gilurytmal, Ignazin, Merabitol, NSC 15627, Raugalline, Rauwolfin, Rauwolfine, Rhytmaton, Ritmos, Rytmalin, Siddiqui, Tachmalin, Takycor |
| InChI Formula | InChI=1S/C20H26N2O2/c1-3-10-11-8-14-17-20(12-6-4-5-7-13(12)21(17)2)9-15(16(11)18(20)23)22(14)19(10)24/h4-7,10-11,14-19,23-24H,3,8-9H2,1-2H3/t10-,11-,14-,15-,16+,17-,18+,19+,20+/m0/s1 |
| SMILES | CC[C@H]1[C@@H]2C[C@H]3[C@@H]4N(C)c5ccccc5[C@]46C[C@@H]([C@H]2[C@H]6O)N3[C@@H]1O |
| IUPAC Name | (1R,9R,10S,12R,13S,14R,16S,17S,18R)-13-ethyl-8-methyl-8,15-diazahexacyclo[14.2.1.01,9.02,7.010,15.012,17]nonadeca-2,4,6-triene-14,18-diol |
| Molecular Weight (g/mol) | 326.43 |
| Formula Weight | 326.1994 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction