missing translation for 'onlineSavingsMsg'
Learn More

Ajmaline, MedChemExpress

Product Code. 16438656
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16438656

Brand: Medchem Express HYB1167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ajmaline (Cardiorythmine) is a sodium channel blocking, class 1A anti-arrhythmic agent. Ajmaline blocks HERG currents with an IC50 of 1 μM in HEK cells and 42.3 μM in Xenopus oocytes. Ajmaline can be used for the research of the ventricular tachyarrhythmia.

Ajmaline (Cardiorythmine) is a sodium channel blocking, class 1A anti-arrhythmic agent. Ajmaline blocks HERG currents with an IC50 of 1 μM in HEK cells and 42.3 μM in Xenopus oocytes. Ajmaline can be used for the research of the ventricular tachyarrhythmia.

Specifications

Chemical Name or Material Ajmaline
CAS 12-7-4360
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H26N2O2
Quantity 100 mg
Synonym Cardiorythmine (+)-Ajmaline
Solubility Information DMSO : ≥ 100 mg/mL (306.34 mM)
SMILES CN1C2=CC=CC=C2[C@@]34[C@]1([H])[C@@](C[C@H]5[C@H](CC)[C@H]6O)([H])[N@]6[C@]([H])([C@H]5[C@H]4O)C3
Molecular Weight (g/mol) 326.43
Formula Weight 326.43
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.