missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AJAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113516
This item is not returnable.
View return policy
Description
AJAP1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
AJAP1, Adherens Junction Associated Protein 1, plays a key role in cell adhesion and cell migration. It is a transmembrane protein found on the basolateral surface of epithelium and expressed by multiple types of epithelium. AJAP1 interacts with E-Cadherin and appears to promote the internalization of E-Cadherin upon growth factor stimulation. This facilitates the dissolution of adherens junctions with an increase in cell motility.
Specifications
| AJAP1 | |
| Polyclonal | |
| Unconjugated | |
| AJAP1 | |
| adherens junction associated protein 1; adherens junction-associated protein 1; adherens junctions associated protein 1; AJAP1; Gm573; likely ortholog of H. sapiens transmembrane protein SHREW1; membrane protein shrew-1; MOT8; Neumo48; RP3426F10\.1; SHREW1; SHREW-1; transmembrane protein SHREW1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 230959, 55966 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| A2ALI5, Q9UKB5 | |
| AJAP1 | |
| Recombinant Human Adherens junction-associated protein 1 protein (308-394AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction