missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Airway Trypsin-like Protease/HAT/TMPRSS11D Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£385.00 - £587.00
Specifications
| Antigen | Airway Trypsin-like Protease/HAT/TMPRSS11D |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18363524
|
Novus Biologicals
NBP3-17648-25UL |
25 μg |
£385.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18329176
|
Novus Biologicals
NBP3-17648-100UL |
100 μg |
£587.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Airway Trypsin-like Protease/HAT/TMPRSS11D Polyclonal antibody specifically detects Airway Trypsin-like Protease/HAT/TMPRSS11D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Airway Trypsin-like Protease/HAT/TMPRSS11D | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| airway trypsin like protease, Airway trypsin-like protease, EC 3.4.21, EC 3.4.21.-, HAT, MGC150587, MGC150588, transmembrane protease serine 11D, transmembrane protease, serine 11D | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: NSNPRDWIATSGISTTFPKLRMRVRNILIHNNYKSATHENDIALVRLENSVTFTKDIHSVCLPAATQNIPPGSTAYVTGWGAQEYAGHTVPELRQGQVRIISNDVCNAPHSYNGAILSGMLCAGV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 9407 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title