Learn More
Hamilton AirShield
Product Code. 31257782
Change view
Click to view available options
For Use With (Equipment):
Microlab 600 Instrument
Product Code. For Use With (Equipment)
31257782 Microlab 600 Instrument
1 options
Product Code. 31257782 Supplier Hamilton Supplier No. 6856201
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AirShield accessory designed for integration with compatible laboratory systems. Supports controlled operation and protection during use.
  • Airshield is an optional accessory for Microlab 600 instruments
  • Creates a positive-pressure airflow inside the pump
  • Directs air over critical components and out of the system
  • Helps protect internal components from environmental contamination
Specifications
Product Type AirShield
For Use With (Equipment) Microlab 600 Instrument