Learn More
Invitrogen™ AIRE Monoclonal Antibody (5H12), eBioscience™, Invitrogen™
Description
Description: This 5H12 monoclonal antibody reacts with mouse Autoimmune Regulator (AIRE). This 58-kDa transcription factor is highly expressed within the perinuclear region of mature medullary thymic epithelial cells (mTECs). However, expression of AIRE protein in the periphery remains unclear despite the detection of AIRE mRNA in the lymph node, lymphoid stromal cells, monocytes, and dendritic cells. Studies demonstrate that AIRE is involved in thymocyte development, in particular the regulation of tissue-specific antigen expression during negative selection. Other biological functions for AIRE include chemokine expression and antigen presentation. The 5H12 antibody recognizes mouse AIRE isoforms 1 and 2. Applications Reported: This 5H12 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, western blotting, and immunohistochemical staining of frozen tissue sections. Applications Tested: This 5H12 antibody has been tested by immunohistochemistry on frozen 5-week-old mouse thymus and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | AIRE |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5H12 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Aire |
| Gene Accession No. | Q9Z0E3 |
| Gene Alias | Aire; AIRE protein; AIRE1; APECED; APECED protein; APECED protein homolog; APS1; APSI; Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein; autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein homolog; Autoimmune regulator; autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy); autoimmune regulator AIRE1a; LOW QUALITY PROTEIN: autoimmune regulator; OTTHUMP00000109529; PGA1 |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.