Learn More
Syngene™ AirClean™ Replacement UV Shelf with Pipette Holder
Use AirClean™ Replacement UV shelf with Pipette Holder for the easy storage of pipettes and pipette tip boxes within a PCR workstation enclosure.
£565.00 - £620.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17735914
|
Syngene™
SYN-ACA1040 |
61 cm (24 in.) and 81 cm (32 in.) Wide PCR Workstation Enclosures |
£565.00
Each |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17745914
|
Syngene™
SYN-ACA1040A |
122 cm (48 in.) Wide PCR Workstation Enclosures |
£620.00
Each |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For the easy storage of pipettes and pipette tip boxes within a PCR workstation enclosure
Specifications
Pipette Holder | |
UV Shelf |