Learn More
Description
AirClean™ Systems manufactures gas phase bonded carbon and multi-layered filters that are available with standard coconut shell carbon or chemically impregnated carbon, depending on your application. AirClean™ Systems gas-phase, coconut shell carbon filters utilize our unique bonded carbon filtration media. This minimizes the release of potentially hazardous carbon dust found in traditional granular carbon filters. Our proprietary bonding process holds carbon in a predictable matrix, preventing carbon shift that can lead to 'dead spots', common in traditional granular filters.
Features
- Bonded carbon — no filter dusting
- Even distribution of impregnates, if applicable
- Multi-layering options for larger application compatibility
Specifications
Specifications
| Filter Type | Carbon |
| Length (English) | 12 in. |
| Depth (English) | 3 in. |
| Width (English) | 24 in. |
| Quantity | 1 Ea. |
| Depth (Metric) | 8 cm |
| Length (Metric) | 30.5 cm |
| Width (Metric) | 61 cm |
| For Use With (Application) | Ammonia, Amines |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
