missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ AirClean™ Blended Bonded Carbon Filter for Acids and Ammonia
GREENER_CHOICE

Product Code. 17755964 Shop All Syngene Products
Change view
Click to view available options
Width (Metric):
46 cm
61 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
17755964 61 cm Each
17765964 46 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17755964 Supplier Syngene™ Supplier No. SYNACF434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The AirClean™ Blended Bonded Carbon Filter for Acids and Ammonia is used to absorb fumes and odors.

AirClean™ Systems manufactures gas phase bonded carbon and multi-layered filters that are available with standard coconut shell carbon or chemically impregnated carbon, depending on your application. AirClean™ Systems gas-phase, coconut shell carbon filters utilize our unique bonded carbon filtration media. This minimizes the release of potentially hazardous carbon dust found in traditional granular carbon filters. Our proprietary bonding process holds carbon in a predictable matrix, preventing carbon shift that can lead to 'dead spots', common in traditional granular filters.

Features

  • Bonded carbon — no filter dusting
  • Even distribution of impregnates, if applicable
  • Multi-layering options for larger application compatibility

Specifications

Filter Type Carbon
Length (English) 12 in.
Depth (English) 3 in.
Width (English) 24 in.
Quantity 1 Ea.
Depth (Metric) 8 cm
Length (Metric) 30.5 cm
Width (Metric) 61 cm
For Use With (Application) Acids, Ammonia
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.