Learn More
Syngene™ Airclean 600 Ductless Workstation, HEPA
Ductless workstation equipped with a HEPA filter and AirSafe controller.
Brand:  Syngene™ SYN-32DWASH-E
Additional Details : Weight : 90.00000kg
Product Code. 16594331
  • £6080.00 / Each
Estimated Shipment: 11-07-2024
to see stock.
Description
Description
  • Airclean 600
  • Ductless
  • With AirSafe controller
  • HEPA filter installed
Specifications
Specifications
HEPA
AirSafe Controller
32 in.
81.28 cm