missing translation for 'onlineSavingsMsg'
Learn More

Draeger™ Air Current Tube Kit

Product Code. 10747674
Change view
Click to view available options
For Use With (Application):
HVAC and mining industries
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
10747674 HVAC and mining industries Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10747674 Supplier Draeger™ Supplier No. CH00216
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in HVAC and mining industries.

  • Reagent inside smoke tube produces visible aerosol plume when in contact with atmospheric humidity
  • Squeeze bulb pushes air through tube to propel smoke into ambient air

Specifications

Type Air Current Tube Kit
For Use With (Application) HVAC and mining industries
compliance-icons
Product Identifier
  • Air current tubes; Strömungsprüfröhrchen
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
  • P402-Store in a dry place.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P420-Store away from other materials.
  • P501-Dispose of contents/container to properly dispose andrecycle.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric acid >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.