Learn More
Moldex™ Air FFP Mask
Brand:  Moldex™ 350501
Product Code. 12312299
Description
Description
Moldex AIR Plus 3505.FFP3 NR D valved dust mask. The unique pleated filter technology offers extremely low breathing resistance. FFP3 with full face seal.