Learn More
Abeba™ Air-Cushion Sole 2700 Shoes
Product Code. 15628247
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15628247
Brand: Abeba™ 2700/47
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Antistatic with air cushion PU-sole and high slip resistance. Sanitized™ treated.
  • CE EN ISO 20347:2012 O2 SRA
  • Grained leather
  • White
  • Slip resistance PU outsole sole
  • Replaceable insole (article no. 3551)
  • Sanitized treated
  • Antistatic
  • Lining microfiber, respiration absorbing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O2 SRA
Color White
Liner Material Microfiber
Material Leather
Size 47
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2700