missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15108-1-AP-20UL
This item is not returnable.
View return policy
Description
Aipl1 was initially identified as a protein implicated in Leber congenital amaurosis (LCA), an autosomal recessive disorder thought to be caused by the abnormal development of photoreceptors. Aipl1 is a tetratricopeptide repeat protein that is highly homologous to ARA9, a protein involved in the HSP90-mediated nuclear translocation and transactivation of the aryl hydrocarbon receptor. Aipl1 has also been found to function as part of a chaperone heterocomplex, interacting with Hsp90 and Hsp70. Aipl1 also associates with the cell cycle regulator NUB1. It is thought that Aipl1 cooperates with Hsp70 but not Hsp90 to suppress the formation of NUB1 inclusions, and these interactions are necessary in the normal photoreceptor maturation, as mutations that lead to LCA also compromise the interactions with the Hsp chaperones. At least three isoforms of Aipl1 are known to exist.Specifications
| AIPL1 | |
| Polyclonal | |
| Unconjugated | |
| AIPL1 | |
| AIPL1, AIPL2, LCA4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114230, 23746, 59110 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q924K1, Q9JLG9, Q9NZN9 | |
| Aipl1 | |
| AIPL1 Fusion Protein Ag4625 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction