missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AIP2 Monoclonal Antibody (K9N015_19D11), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | AIP2 |
|---|---|
| Clone | K9N015_19D11 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30202947
|
Invitrogen™
MA556079 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30210606
|
Invitrogen™
MA555699 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: TAVAPENRHQ PPSTNCFGGR SRTHRHSGAS ARTTPATGEQ SPGARSRHRQ PVKNSGHSGL ANGTVNDEPT TATDPEEPSV VGVTSPPAAP LSVTPNPNTT SLPAPATPAE GEEPSTSGTQ QLPAAAQAPD ALPAGWEQRE LPNGRVYYVD HNTKTTTWER PLPPGWEKRT DPRGRFYYVD HNTRTTTWQR PTAEYVRNYE
AIP2 Monoclonal antibody specifically detects AIP2 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| AIP2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1300010O06Rik; AA690238; AIP2; atrophin-1 interacting protein 2; Atrophin-1-interacting protein 2; AW554328; HECT-type E3 ubiquitin transferase WWP2; id:ibd1121; NEDD4-like E3 ubiquitin-protein ligase WWP2; wu:fo87e10; WW domain containing E3 ubiquitin protein ligase 2; WW domain-containing protein 2; WW domain-containing protein 4; WWP2; WWp2-like; zgc:154036 | |
| Recombinant human WWP2 (immunogen range is 171-370). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| WWP2 |
| K9N015_19D11 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O00308 | |
| 11060 | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title