missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AIP2 Monoclonal Antibody (K9N015_19D11), Invitrogen™

Product Code. 30202947
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30202947 200 μL 200µg
30210606 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30202947 Supplier Invitrogen™ Supplier No. MA556079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: TAVAPENRHQ PPSTNCFGGR SRTHRHSGAS ARTTPATGEQ SPGARSRHRQ PVKNSGHSGL ANGTVNDEPT TATDPEEPSV VGVTSPPAAP LSVTPNPNTT SLPAPATPAE GEEPSTSGTQ QLPAAAQAPD ALPAGWEQRE LPNGRVYYVD HNTKTTTWER PLPPGWEKRT DPRGRFYYVD HNTRTTTWQR PTAEYVRNYE

This gene encodes a member of the NEDD4-like protein family. The family of proteins is known to possess ubiquitin-protein ligase activity. The encoded protein contains 4 tandem WW domains. The WW domain is a protein motif consisting of 35 to 40 amino acids and is characterized by 4 conserved aromatic residues. The WW domain may mediate specific protein-protein interactions. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIP2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K9N015_19D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene WWP2
Gene Accession No. O00308
Gene Alias 1300010O06Rik; AA690238; AIP2; atrophin-1 interacting protein 2; Atrophin-1-interacting protein 2; AW554328; HECT-type E3 ubiquitin transferase WWP2; id:ibd1121; NEDD4-like E3 ubiquitin-protein ligase WWP2; wu:fo87e10; WW domain containing E3 ubiquitin protein ligase 2; WW domain-containing protein 2; WW domain-containing protein 4; WWP2; WWp2-like; zgc:154036
Gene Symbols WWP2
Host Species Mouse
Immunogen Recombinant human WWP2 (immunogen range is 171-370).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11060
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.