Learn More
Invitrogen™ AIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114129
Description
Immunogen sequence: FHYRTLHSDD EGTVLDDSRA RGKPMELIIG KKFKLPVWET IVCTMREGEI For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 79%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Rat: 86%.
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
Specifications
| AIP | |
| Polyclonal | |
| Unconjugated | |
| Aip | |
| AA408703; AH receptor-interacting protein; AIP; ARA9; aryl hydrocarbon receptor interacting protein; aryl-hydrocarbon receptor-interacting protein; AW476050; D19Bwg1412e; FKBP16; FKBP37; HBV X-associated protein 2; HBV-X associated protein; immunophilin homolog ARA9; Immunophilin XAP2; SMTPHN; XAP2; XAP-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9049 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00170 | |
| Aip | |
| Synthetic peptide directed towards the N terminal region of human AIP (aa 36-85). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.