missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AIP Polyclonal Antibody

Product Code. 13454197
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13454197 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13454197 Supplier Invitrogen™ Supplier No. PA529862

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: Molt-4. Predicted reactivity: Human (99%), Mouse (94%), Rat (93%), Chicken (82%), Rhesus Monkey (97%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Aip
Gene Accession No. O00170, O08915
Gene Alias AA408703; AH receptor-interacting protein; AIP; ARA9; aryl hydrocarbon receptor interacting protein; aryl-hydrocarbon receptor-interacting protein; AW476050; D19Bwg1412e; FKBP16; FKBP37; HBV X-associated protein 2; HBV-X associated protein; immunophilin homolog ARA9; Immunophilin XAP2; SMTPHN; XAP2; XAP-2
Gene Symbols Aip
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ARA9. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11632, 9049
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.